Learn More
ACROBiosystems ACROBiosystems Human PLAU / uPA Protein, His Tag (activated by trypsin)
Supplier:  ACROBiosystems PLUH522850UG
Encompass_Preferred
Recombinant Protein;50UG;Human PLAU, His Tag (activated by trypsin) (PLU-H5228) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431  (Accession # P00749-1).;This protein carries a polyhistidine tag at the C-terminus. The active form of Human PLAU is a disulfide-linked heterodimer composed of long chain A (Ser 21 - Phe 177) and chain B (Ile 179 - Leu 431) with calculated MW of 17.8 kDa and 29.2 kDa. The long chain A is further cleaved to yield a short chain A (Lys 156 - Phe 177) and N-terminal fragment (Ser 21 - Lys 155) with calculated MW of 15.3 kDa. The protein migrates as 17 kDa (N-terminal fragment), 32-35 kDa (chain B) and 45-50 kDa (long chain A & chain B) under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6717
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.