Learn More
Invitrogen™ PLBD1 Polyclonal Antibody

Description
PLBD1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PLBD1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PLBD1 |
| Gene Accession No. | Q6P4A8 |
| Gene Alias | 1100001H23Rik; hypothetical protein; im:7138859; LAMA-like protein 1; Lamina ancestor homolog 1; phospholipase B domain containing 1; phospholipase B domain-containing protein 1; Phospholipase B-like 1; Phospholipase B-like 1 chain A; Phospholipase B-like 1 chain B; Phospholipase B-like 1 chain C; PLB homolog 1; PLBD1; putative phospholipase B-like 1; RGD1308734; zgc:194816 |
| Gene Symbols | PLBD1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.