Learn More
MyBioSource Phospholipase B-like 2 ELISA Kit
SDP
Supplier:  MyBioSource MBS5642021 KIT
Encompass_Preferred
The MyBioSource Phospholipase B-Like 2 (PLBL2) ELISA Kit quantitatively measures human PLBL2 in various biological fluids using a 96-well plate coated with a specific antibody. After binding and washing steps, a biotinylated detection antibody and HRP-conjugated streptavidin are added. TMB substrate reveals the PLBL2 level by a color change measured at 450 nm.
Catalog No. NC1486072
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.