Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCB3 Mouse anti-Human, Mouse, Rat, Clone: 1B8B3, Proteintech
SDP

Catalog No. 501737085 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7085 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7085 Supplier Proteintech Group Inc Supplier No. 666681IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.

Specifications

Antigen PLCB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1B8B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PLCB3
Gene Accession No. P51432, Q01970, Q99JE6
Gene Alias FLJ37084, Phospholipase C beta 3, PLC beta 3, PLCB3
Gene Symbols Plcb3
Host Species Mouse
Immunogen PLCB3 Fusion Protein Ag15845
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18797, 29322, 5331
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.