Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCXD1 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
SDP

Catalog No. p-200016907
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2356 Supplier OriGene Supplier No. TA808765
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is the most terminal protein-coding gene in the pseudoautosomal (PAR) region on chromosomes X and Y. Alternate splicing results in multiple transcript variants.

Specifications

Antigen PLCXD1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI2D7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLCXD1
Gene Accession No. Q9NUJ7
Gene Alias LL0XNC01-136G2.1
Gene Symbols PLCXD1
Host Species Mouse
Immunogen Full length human recombit protein of human PLCXD1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55344
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.