Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCXD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571235 Supplier Invitrogen™ Supplier No. PA571235
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

PLCXD3 gene ontology annotations related to this gene include lipid metabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCXD3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLCXD3
Gene Accession No. Q63HM9
Gene Alias B130016O10Rik; phosphatidylinositol specific phospholipase C X domain containing 3; phosphatidylinositol-specific phospholipase C, X domain containing 3; PI-PLC X domain-containing protein 3; PLCXD3; RGD1308448
Gene Symbols PLCXD3
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human PLCXD3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 345557
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.