Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598556
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598556 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598556 Supplier Invitrogen™ Supplier No. PA598556
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLCZ1 Polyclonal Antibody for ELISA, Western Blot

The protein encoded by this gene is a member of the phosphoinositide-specific phospholipase C family. Members in this family, classified into six isotypes that are tissue- and organ-specific, hydrolyze phosphatidylinositol 4,5-bisphosphate just before the phosphate group to yield diacylglycerol and inositol 1,4,5-trisphosphate. This protein localizes to the acrosome in spermatozoa and elicits Ca(2+) oscillations and egg activation during fertilization that leads to early embryonic development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCZ1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Plcz1
Gene Accession No. Q86YW0, Q8K4D7
Gene Alias 1700041H07Rik; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase zeta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase zeta-1; NYD-SP27; Phosphoinositide phospholipase C-zeta-1; phospholipase C zeta; phospholipase C zeta 1; phospholipase C, zeta 1; phospholipase C-zeta-1; phospholipase C-zeta-1 {ECO:0000250; PI-phospholipase C zeta 1; PLCZ1; plcz1 {ECO:0000312; PLCzeta; PLC-zeta-1; PLC-zeta-1 {ECO:0000250; RGD:1359567}; testicular tissue protein Li 145; Testis-development protein NYD-SP27; testis-development related NYD-SP27; UniProtKB:Q86YW0}
Gene Symbols Plcz1
Host Species Rabbit
Immunogen Recombinant Human 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase zeta-1 protein (1-415AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114875, 89869
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.