Learn More
Bachem Biotinyl-ACTH (1-39) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4033441.0500
Encompass_Preferred
Biotinyl-ACTH (1-39) (human) Trifluoroacetate, Biotinyl-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-Asn-Gly-Ala-Glu-Asp-Glu-Ser-Ala-Glu-Ala-Phe-Pro-Leu-Glu-Phe-OH, 1816258-26-0, C217H322N58O60S2, Mr 4767.43, 0.5mg. Biotin-ACTH (1-39), Human. ACTH (1–39) is a member of the melanocortin receptor agonists, which stimulates the adrenal cortex and the secretion of glucocorticoids such as cortisol.
Catalog No. 50-194-6692