Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLEKHA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548911
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548911 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548911 Supplier Invitrogen™ Supplier No. PA548911
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLEKHA6 Polyclonal Antibody for Western Blot

PLEKHA6 (Pleckstrin Homology Domain Containing A6) is a Protein Coding gene. Among its related pathways arePI MetabolismandGlycerophospholipid biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLEKHA6
Applications Western Blot
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PLEKHA6
Gene Accession No. Q9Y2H5
Gene Alias 9530028L01Rik; AU042621; KIAA0969; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family A member 6; PEPP3; PEPP-3; PH domain-containing family A member 6; phosphoinositol 3-phosphate-binding protein 3; phosphoinositol 3-phosphate-binding protein-3; pleckstrin homology domain containing A6; pleckstrin homology domain containing, family A member 6; pleckstrin homology domain-containing family A member 6; pleckstrin homology domain-containing family A member 6; LOW QUALITY PROTEIN: pleckstrin homology domain-containing family A member 6; Plekha6; RGD1564395
Gene Symbols PLEKHA6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 710-736 amino acids from the Central region of human PLEKHA6.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.