Learn More
Selleck Chemical LLC Plerixafor (AMD3100) 5mg 110078-46-1 JM 3100
SDP
Supplier:  Selleck Chemical LLC S80305MG
Encompass_Preferred
Plerixafor (AMD3100, JM 3100) is a chemokine receptor antagonist for CXCR4 and CXCL12-mediated chemotaxis with IC50 of 44 nM and 5.7 nM in cell-free assays, respectively. Plerixafor inhibits human immunodeficiency virus (HIV) replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5581
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.