Learn More
Selleck Chemical LLC Pleuromutilin S4436-5mg
SDP
Supplier:  Selleck Chemical LLC S44365MG
Encompass_Preferred
Pleuromutilin (Drosophilin B Mutilin 14-glycolate) the lead compound for novel antibiotics inhibits bacterial protein synthesis by binding to the 50S ribosomal subunit of bacteria
Catalog No. 50-311-0198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.