Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Plexin A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543457 Supplier Invitrogen™ Supplier No. PA543457
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SVASYVYNGY SVVFVGTKSG KLKKIRADGP PHGGVQYEMV SVLKDGSPIL Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Plexins are a family of large, transmembrane receptors for multiple classes of semaphorins in vertebrates. Plexins are widely expressed, and regions of their extracellular domain are homologus to both scatter factor receptors and semaphorin domains. Plexins may act as semaphorin receptors alone or in combination with neuropilins. Plexins are divided into four subfamilies designated plexin-A, -B, -C, and -D. Drosophila plexin-A is a receptor for class I semaphorins and controls motor and axon guidance. Plexin-A3 mediates cell-repelling cues. Plexins B and C are receptors for Sema 4 and Sema 7, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PLXNA2
Gene Accession No. O75051
Gene Alias 2810428A13Rik; AA589422; AW457381; FLJ11751; FLJ30634; KIAA0463; LOW QUALITY PROTEIN: plexin-A2; mKIAA0463; OCT; plex 2; PlexA2; plexin 2; plexin A2; plexin-2; plexin-A2; PLXN2; Plxna2; Semaphorin receptor OCT; si:dkey-164k14.1; transmembrane protein OCT; UNQ209/PRO235
Gene Symbols PLXNA2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PLXNA2 (aa 434-483).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.