Learn More
ECM Bioscience Plexin A1 (Sema Domain) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PP1301
Encompass_Preferred
The antibody detects a 220 kDa* protein corresponding to the apparent molecular mass of Plexin-A1 on SDS-PAGE immunoblots of neonatal rat brain and Cos-7 cells transfected with mouse Plexin-A1. In immunocytochemistry, this antibody co-localizes with anti-Myc in Cos-7 cells transfected with Myc-tagged mouse Plexin-A1. The immunogen was Plexin-A1 synthetic peptide (coupled to carrier protein) corresponding to a region within the N-terminal half of the semaphorin domain in human Plexin-A1. This sequence is highly conserved in rat and mouse Plexin-A1, and has low homology to other Plexin family members. Accession #Q9UIW2. Applications = WB, E, IP, ICC, IHC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = PLXN1, NOV, Sema3A
Catalog No. 50-172-0861
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.