Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLK1 Recombinant Rabbit Monoclonal Antibody (K01_2Q37)

Catalog No. MA565723
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA565723 50 μL
MA564264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA565723 Supplier Invitrogen™ Supplier No. MA565723
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: ERDGTESYLT VSSHPNSLMK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%, Pig - 95%, Cynomolgus monkey - 100%.

PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLK1
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2Q37
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Plk1
Gene Accession No. P53350, Q62673
Gene Alias cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg
Gene Symbols Plk1
Host Species Rabbit
Immunogen A synthetic peptide of human PLK1 (aa 455-474).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25515, 5347
Target Species Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.