Learn More
Clontech Labs 3P PLRES2-ZSGREEN1 VECTOR 20UG
SDP
Supplier:  Clontech Labs 3P 632478
Encompass
pIRES2-ZsGreen1 is a bicistronic expression vector containing ZsGreen1 as a marker for transfection efficiency. It encodes a human-codon-optimized variant of the Zoanthus sp. green fluorescent protein, ZsGreen, which has been engineered for brighter fluorescence and higher expression in mammalian cells. A gene of interest is inserted into the MCS located upstream of the encephalomyocarditis virus (ECMV) internal ribosomal entry site (IRES). The IRES sequence allows the gene of interest and ZsGreen1 to be translated simultaneously from the same mRNA transcript.
Catalog No. NC1864918
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.