Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLSCR2 Monoclonal Antibody (OTI7A5), TrueMAB™, OriGene
SDP

Catalog No. 501683679
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3679 Supplier OriGene Supplier No. TA810855
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.; FUNCTION: Isoform 1 has no prospholipid scramblase activity, due to the lack of a N-terminal proline-rich domain.

Specifications

Antigen PLSCR2
Applications Western Blot
Classification Monoclonal
Clone OTI7A5
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PLSCR2
Gene Accession No. Q9DCW2, Q9NRY7
Gene Alias phospholipid scramblase 2
Gene Symbols Plscr2
Host Species Mouse
Immunogen Full length human recombit protein of human PLSCR2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18828, 57047
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.