Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Pluripotent Stem Cell Transcription Factor Antibody Pack
SDP

Catalog No. NBP142823 Shop All Novus Products
Change view
Click to view available options
Quantity:
3 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP142823 3 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP142823 Supplier R&D Systems™ Supplier No. NBP142823

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Includes

  • 1.NB100-2379: OCT4 Antibody
  • 2.NB100-588: Nanog Antibody
  • 3.NB110-37235: SOX2 Antibody

A Novus product, part of R&D Systems. For Research applications

Pluripotent Stem Cell Transcription Factor Polyclonal specifically detects Pluripotent Stem Cell Transcription Factor in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Pluripotent Stem Cell Transcription Factor
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Host Species Rabbit
Immunogen NB100-58842: A synthetic peptide made to the mouse Nanog protein (within residues 1-50). [Swiss-Prot Q80Z64]. NB100-2379-A synthetic peptide made to an internal portion
Purification Method Affinity Purified
Quantity 3 Vials
Research Discipline Stem Cell Markers
Test Specificity NB100-58842 is specific for mouse Nanog protein. NB100-2379-OCT4 - Embryonic Stem Cell Marker NB110-37235 - SOX2 - Embryonic Stem Cell Marker
Includes 1.NB100-2379: OCT4 Antibody
2.NB100-588: Nanog Antibody
3.NB110-37235: SOX2 Antibody
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.