Learn More
Sigma Aldrich Fine Chemicals Biosciences Pluronic F-127, 9003-11-6, MFCD00082049, 250G
Supplier: Sigma Aldrich Fine Chemicals Biosciences P2443250G
Pluronic F-127 was used to coat a siliconized coverslip to hold an egg extract in a study. Pluronic F-127 was added to phosphate buffered saline (PBS), to lower unspecific cell and protein adhesion to a PDMS-based microfluidic device. A study reports its use as a release vehicle to transport low-dose perivascular lipopolysaccharide (LPS) on mouse vein grafts. PLGA/pluronic F127 may be used to fabricate nerve guidance conduits (NGCs) for regeneration of peripheral nerve. Fabrication of poly (lactide-co-glycolide) (PLGA) — Pluronic F127 glass composites was reported. Pluronic F-127 was used for fluorescent labeling of blood vessels, astrocytes, and neurons.