Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLURONIC™ F-127, Proteing Grade™ Detergent, 10% Solution, Sterile-Filtered, MilliporeSigma™

Catalog No. 54002550ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-002-550ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 54-002-550ML Supplier MilliporeSigma Supplier No. 54002550ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9004-07-3
Synonym Poloxamer 407, Polyoxypropylenepolyoxyethylene Block Copolymer

Specifications

Color Undesignated
Quantity 50 mL
Packaging Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.