Learn More
Sartorius Sartorius iQue Plus One Detector Maintenance Solution, 1X Concentration, Lubricant for Clog Prevention, 5 x 500 ml
Catalog No. NC1321030 Shop All Sartorius Products
Encompass
The Sartorius iQue Plus One Detector Maintenance Solution is a ready-to-use liquid designed for routine maintenance of flow cytometry equipment. This product ensures optimal performance by acting as a lubricant to prevent clogging and facilitate the cleaning process. Its 1X concentration and 5-pack packaging offer convenience and efficiency, making it essential for maintaining detector cleanliness during startup and shutdown cycles.
Catalog No. NC1321030 Supplier Sartorius Supplier No. 91089
The Sartorius iQue Plus One Detector Maintenance Solution is a ready-to-use liquid designed for routine maintenance of flow cytometry equipment. This product ensures optimal performance by acting as a lubricant to prevent clogging and facilitate the cleaning process. Its 1X concentration and 5-pack packaging offer convenience and efficiency, making it essential for maintaining detector cleanliness during startup and shutdown cycles.