Learn More
Abcam (±)-BAYK8644, L-type Ca2+ channel activator, 50MG
Supplier:  Abcam AB12036650MG
Encompass_Preferred
MW 356.3 Da, Purity >99%. Potent L-type Ca²+ channel activator (EC₅₀ = 17.3 nM). Centrally active following systemic administration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7646