Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva PlusOne™ Reagents

Catalog No. 45000234 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 g
500 mL
Form:
Liquid
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
45-000-234 500 g Solid
17-133-201 500 mL Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45-000-234 Supplier Cytiva Supplier No. 17131901
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality electrophoresis reagents. Cytiva Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.

Urea

  • Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
  • Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
  • 8M urea included in the 2-D sample solution
Repel-Silane ES
Low toxicity and environmentally safe solvent
  • Inhibits sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces
  • Bind-Silane
    • Used for covalently attach polyacrylamide gels to a glass surface
    • Firmly attaches to the glass during staining and drying procedures

    Urea:

    • Molecular Weight: 60.06
    • Physical state: Solid
    • Color: White
    • Pack of 500g
    Repel-Silane ES
    • Molecular Weight: 129.06
    • Physical state: Liquid
    • Color: Colorless
    • Pack of 500mL
    Bind-Silane
    • Molecular Weight: 248.4
    • Physical state: Liquid
    • Color: Pale
    • Pack of 25mL

    IEF, PAGE, sequencing

    Certifications

    Suitable for DIGE

    TRUSTED_SUSTAINABILITY

    Specifications

    Boiling Point > 132°C
    CAS 2530-85-0
    Color White
    Form Solid
    For Use With (Application) IEF, PAGE, sequencing
    Melting Temperature 132°C
    Molecular Weight (g/mol) 60.06
    Quantity 500 g
    Storage Requirements Dry at 5 - 30ᵒC
    Product Type Urea
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.