Learn More
Medchemexpress LLC HY-114153 5mg Medchemexpress, PLX5622 CAS:1303420-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1141535MG
Encompass_Preferred
Medchemexpress, HY-114153 5mg PLX5622 CAS:1303420-67-8 PLX5622 is a highly selective brain penetrant and oral active CSF1R inhibitor with an Ki of 5.9 nM, for extended and specific microglial elimination, preceding and during pathology development. PLX5622 demonstrates desirable PK properties in varies animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.