Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566227 Supplier Invitrogen™ Supplier No. PA566227
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSLDAGSRVTV TVRDSECQFV RRDAKAIVCI SPLSTLGPSQ APITLAIDRA NISSPGLIYT YTQDPTVTRL EPTWSIINGS TA Highest antigen sequence identity to the following orthologs - mouse 84%, rat 86%.

Coreceptor for SEMA3A and SEMA3F. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Regulates the migration of sympathetic neurons, but not of neural crest precursors. Required for normal dendrite spine morphology in pyramidal neurons. May play a role in regulating semaphorin-mediated programmed cell death in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNA3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Plxna3
Gene Accession No. P51805
Gene Alias 6.3; HSSEXGENE; mKIAA4078; PlexA3; plexin 3; Plexin 4, SEX, homolog to the cMet/HGF receptors; plexin A3; plexin-4; Plexin-A3; Plxa3; Plxn3; Plxn4; Plxna3; Semaphorin receptor SEX; SEX; Sex chromosome X transmembrane protein of HGF receptor family 3; SEXPLEXIN-A3; XAP-6
Gene Symbols Plxna3
Host Species Rabbit
Immunogen Recombinant Human PLXNA3. Recombinant protein control fragment (Product #RP-105119).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55558
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.