Learn More
Akina Inc Poly(lactide-co-glycolide)-Flamma Fluor FKR648 (per 100mg)
SDP
Supplier:  Akina Inc AV015100MG
Encompass
PLGA-FKR648 FKR648 endcap (Mn: 20000-30000 Da) Poly(lactide-co-glycolide)-Flamma Fluor FKR648 (per 100mg)
Catalog No. NC2827977