Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Monoclonal Antibody (2A9), Active Motif™
SDP

Catalog No. 501993018
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3018 10 μg
50-199-3017 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3018 Supplier Active Motif Supplier No. 39988
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.

Specifications

Antigen PLZF
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene ZBTB16
Gene Accession No. Q05516
Gene Alias AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; Zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); Zinc finger protein PLZF; ZNF145
Gene Symbols ZBTB16
Host Species Mouse
Immunogen This PLZF antibody was raised against a recombinant protein corresponding to full-length mouse PLZF with a GST-Tag.
Purification Method Protein G
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235320, 7704
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.