Learn More
Invitrogen™ PLZF Monoclonal Antibody (5B3)

Description
MA5-15667 targets ZBTB16 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15667 immunogen is purified recombinant fragment of human ZBTB16 expressed in E. Coli.. MA5-15667 detects ZBTB16 which has a predicted molecular weight of approximately 74kDa.
Specifications
Specifications
| Antigen | PLZF |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5B3 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | ZBTB16 |
| Gene Accession No. | Q05516 |
| Gene Alias | AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.