Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Mouse anti-Human, Clone: 2E6F12, Proteintech
SDP

Catalog No. p-7212703 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7089 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7089 Supplier Proteintech Group Inc Supplier No. 666721IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia. Alternate transcriptional splice variants have been characterized.

Specifications

Antigen PLZF
Applications Western Blot
Classification Monoclonal
Clone 2E6F12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZBTB16
Gene Accession No. Q05516
Gene Alias PLZF, ZBTB16, Zinc finger protein 145, Zinc finger protein PLZF, ZNF145
Gene Symbols ZBTB16
Host Species Mouse
Immunogen PLZF Fusion Protein Ag25180
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7704
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.