Learn More
Description
Immunogen sequence: HYTLDFLSPK TFQQILEYAY TATLQAKAED LDDLLYAAEI LEIEYLEEQC LKMLETIQAS DDNDTEATMA DGGAEEEEDR KARYLKNIFI SKHSSEESGY ASVAGQSLPG PMVDQSPSVS TSFGLSA Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 97%.
Specifications
Specifications
| Antigen | PLZF |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZBTB16 |
| Gene Accession No. | Q05516 |
| Gene Alias | AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 |
| Gene Symbols | ZBTB16 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
