Learn More
Warner Instruments™ PM-1 Platform For Series 20 Chambers, Heated, Magnetic Clamps
Catalog No. BTX641526 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Series 20 Chambers
Catalog No. For Use With (Equipment)
BTX641526 Series 20 Chambers
1 options
Catalog No. BTX641526 Supplier Warner Instruments™ Supplier No. 641526
Heated magnetic platform provides a base for Series 20 chambers and clamps the chamber to the coverslip to form a seal.
  • Allows small entry angles suitable for patch recordings
  • No tools needed to secure chamber to platform
  • Available for most Series 20 Chambers
  • Magnetic stainless steel allows use of magnetic holders
  • Compatible with all series 20 stage adapters
  • Platforms come with heating elements
Specifications
For Use With (Equipment) Series 20 Chambers