Learn More
Thermo Scientific Dionex Aquion Preventive Maintenance Kit | | 1 EA
Supplier: Thermo Scientific 057954
Preventive maintenance manual, O-ring for pump spacer 2, Eluent end-line filter 1, Check valve cartridge 2, Pump rinse seal 2, O-ring for waste and priming valves 2, Piston seal 2, O-ring for pump piston rinse seal 2, 6-port Injection valve rebuild kit 1, Reminder label for next PM