Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PM20D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546590 Supplier Invitrogen™ Supplier No. PA546590
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SMGPRSGEHQ RASRIPSQFS KEERVAMKEA LKGAIQIPTV TFSSEKSNTT Sequence homology: Guinea Pig: 91%; Human: 100%.

Bidirectional N-fatty-acyl amino acid synthase/hydrolase that regulates the production of N-fatty-acyl amino acids. These metabolites are endogenous chemical uncouplers of mitochondrial respiration. In an UCP1-independent manner, maybe through interaction with mitochondrial transporters, they promote proton leakage into the mitochondrial matrix. Thereby, this secreted protein may indirectly regulate the bodily dissipation of chemical energy as heat through thermogenic respiration. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PM20D1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Pm20d1
Gene Accession No. Q6GTS8
Gene Alias Cps1; N-fatty-acyl-amino acid synthase/hydrolase PM20D1; peptidase M20 domain containing 1; peptidase M20 domain-containing protein 1; PM20D1; probable carboxypeptidase PM20D1
Gene Symbols Pm20d1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PM20D1 (aa 25-74).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148811
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.