Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMA1 Monoclonal Antibody (40B7)
GREENER_CHOICE

Catalog No. MA191567 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA191567 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA191567 Supplier Invitrogen™ Supplier No. MA191567
  • $642.00
    $506.65 / Each of 1

    Save $135.35

    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

The predicted molecular weight by western blot is 100 kDa.

The PMA1 (Plasma Membrane ATPase 1) gene encodes a highly conserved enzyme that functions as a proton pump (H+ ATPase) in the plasma membrane of yeast and plant cells. This enzyme is essential for maintaining cellular pH and membrane potential by actively transporting protons (H+) out of the cell, using energy derived from ATP hydrolysis. In Saccharomyces cerevisiae (baker's yeast), PMA1 is vital for nutrient uptake, cell growth, and overall cellular homeostasis. The activity of PMA1 is regulated through various mechanisms, including phosphorylation and interaction with regulatory proteins. In plants, homologs of PMA1 play a crucial role in processes like stomatal opening, nutrient uptake, and stress responses. Given its fundamental role in cellular physiology, PMA1 has been a target for antifungal agents and herbicides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMA1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 40B7
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Tissue culture supernatant with 10mM sodium azide
Gene PMA1
Gene Accession No. P05030
Gene Alias H(+)-exporting P2-type ATPase PMA1; KTI10; Plasma membrane ATPase 1; PMA1; Proton pump 1; YGL008C
Gene Symbols PMA1
Host Species Mouse
Immunogen Yeast membrane preps.
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 852876
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.