Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117327
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117327 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117327 Supplier Invitrogen™ Supplier No. PA5117327
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

MCH may act as a neurotransmitter or neuromodulator in a broad array of neuronal functions directed toward the regulation of goal-directed behavior, such as food intake, and general arousal. May also have a role in spermatocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCH
Applications Western Blot
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PMCH
Gene Accession No. P14200, P20382, P56942
Gene Alias A230109K23Rik; MCH; melanin-concentrating hormone; melanin-concentrating hormone precursor; NEI; Neuropeptide-glutamic acid-isoleucine; neuropeptide-glutamic acid-isoleucine (NEI); Neuropeptide-glycine-glutamic acid; neuropeptide-glycine-glutamic acid (NGE); NGE; Pmch; ppMCH; prepro-MCH; prepro-melanin-concentrating hormone; pro-MCH; pro-melanin concentrating hormone; pro-melanin-concentrating hormone
Gene Symbols PMCH
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PMCH. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110312, 24659, 5367
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.