Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell BW™ Clip™ 3-year Hydrogen Sulfide Single Gas Monitor

Catalog No. 19817106
Encompass
Change view
Click to view available options
Components Measured:
Hydrogen Sulfide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-817-106 Hydrogen Sulfide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-817-106 Supplier Honeywell Supplier No. BWC3H

Please call customer service for current pricing. 1-800-766-7000

BW Clip Series single-gas detector is an everyday companion for hazardous environments

  • Operates maintenance-free: Just turn on the device and it runs continuously — no need for calibration, sensor replacement, battery replacement or charging
  • Compact, lightweight design with one-button operation
  • Designed for a range of harsh environments and extreme temperatures
  • Hibernation mode with case accessory or IntelliDoX
  • Automated self-test of battery, sensor and electronics
  • Wide-angle flash, which alerts simultaneously with audible and vibrating alarm
  • Automatic logging of the 35 most recent gas events and bump test results
  • Affordable, with low cost of ownership
  • Device Management with Honeywell SafetySuite
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance UL 913 8th Edition UL 60079-0:2013, UL 60079-11:2013 CSA C22.2 No. 157-92:2012, CSA C22.2 No. 60079-0:2011, CSA C22.2 No. 60079-11:2014 EN 60079-0:2012 +A11:2013, EN 60079-11:2012 EN 60079-26:2007 IEC 60079-0:2011 IEC 60079-11:2011 IEC 60079-26:2006
Color Yellow
Components Measured Hydrogen Sulfide
Dimensions (L x W x H) 1.1 x 2.0 x 3.4 in. / 2.8 x 5.0 x 8.7 cm
Type Single-gas Detector
For Use With (Application) Single-gas Detector
Height (Metric) 8.7 cm
Includes Standard Package Contents: Detector complete with specified sensor, stainless steel alligator clip and concussion-proof housing; Test cap; Instructions
Length (Metric) 2.8 cm
Material Polycarbonate substrate interior and (b) a thermoplastic elastomer conductive overmold
Weight (Metric) 92 g
Width (Metric) 5.0 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.