Learn More
United Scientific™ PMP Graduated Beaker Set
SDPSCI_ED
Catalog No. S43713 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
PMP
Catalog No. Material
S43713 PMP
1 options
Catalog No. S43713 Supplier United Scientific™ Supplier No. BMSET5
Add to Cart
Add to Cart
This PMP Graduated Beaker Set includes 5 beakers in sizes ranging from 50 mL to 1000 mL.
  • Polymethylpentene offers glass-like clarity with the durability of plastic.
  • Set includes 5 graduated beakers.
  • Beaker Volumes: 50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL
Specifications
Color Clear
Capacity (Metric) 50 mL, 100 mL, 250 mL, 500 mL, and 1000 mL
Material PMP
Graduation Range Various
Includes 5 Beakers
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Molded Graduations
Style Griffin, Low Form