Learn More
Invitrogen™ PMS2 Monoclonal Antibody (163C1251)

Description
Suggested positive control: NIH-3T3. Immunogen sequence: SSLAKRIKQL HHEAQQS.
Specifications
Specifications
| Antigen | PMS2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 163C1251 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | PMS2 |
| Gene Accession No. | P54278, P54279 |
| Gene Alias | AW555130; DNA mismatch repair protein PMS2; H_DJ0042M02.9; HNPCC4; mismatch repair endonuclease PMS2; MLH4; PMS1 homolog 2, mismatch repair protein; PMS1 homolog 2, mismatch repair system component; PMS1 protein homolog 2; PMS2; PMS2 postmeiotic segregation increased 2; PMS2 postmeiotic segregation increased 2 (S. cerevisiae); PMS2CL; PMSL2; postmeiotic segregation increased 2; postmeiotic segregation increased 2 (S. cerevisiae); postmeiotic segregation increased 2 nirs variant 6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.