Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam PMSF Protease Inhibitor, Solid, >98% Purity, 100 g

Catalog No. 502583377
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3377 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3377 Supplier Abcam Supplier No. AB141032100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-specific protease inhibitor that binds to serine residues and crosses the blood-brain barrier.

  • Molecular Weight: 174.19 Da
  • Purity: >98%
  • Physical State: Solid
  • Function: Non-specific protease inhibitor; binds active site serine residues in serine proteases, acetylcholinesterases, and thiol proteases
  • Applications: Suitable for use in vitro, preparation of cell lysates, and in vivo studies
  • Features: Rapidly crosses the blood-brain barrier and blocks anandamide metabolism
  • Composition: 99.19% PMSF

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Serine Proteases, Acetylcholinesterases, Thiol Proteases
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Buffer 99.19% PMSF
Content And Storage Store at -20°C.
Form Solid
Formulation 99.19% PMSF
Quantity 100 g
For Use With (Application) In vitro use, preparation of cell lysates, in vivo block anandamide metabolism
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.