Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMVK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579838
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579838 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579838 Supplier Invitrogen™ Supplier No. PA579838
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Cardiac Muscle Tissue. IHC: Rat Skeletal Muscle Tissue.

PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMVK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Pmvk
Gene Accession No. Q15126, Q9D1G2
Gene Alias 1110011E12Rik; 2900002L22Rik; AV001327; hPMK; HUMPMKI; Phosphomevalonate kinase; PMK; PMKA; PMKASE; PMKI; Pmvk; POROK1; testis tissue sperm-binding protein Li 95mP
Gene Symbols Pmvk
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human PMVK (135-151aa EQSRQQRGWVFTPGVDD).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10654, 310645, 68603
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.