Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMVK Recombinant Rabbit Monoclonal Antibody (014)
GREENER_CHOICE

Catalog No. PIMA529488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529488 Supplier Thermo Scientific Supplier No. MA529488
Only null left
Add to Cart
Add to Cart

PMVK Recombinant Rabbit Monoclonal Antibody (14)

PMVK Recombinant Monoclonal Antibody for Western Blot, IP, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PMVK
Applications ELISA, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone 014
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Pmvk
Gene Accession No. Q15126
Gene Alias 1110011E12Rik; 2900002L22Rik; AV001327; hPMK; HUMPMKI; Phosphomevalonate kinase; PMK; PMKA; PMKASE; PMKI; Pmvk; POROK1; testis tissue sperm-binding protein Li 95mP
Gene Symbols Pmvk
Host Species Rabbit
Immunogen Recombinant Human PMVK/phosphomevalonate kinase Protein (Met1-Leu192).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.