Learn More
Medchemexpress LLC Ac-Phe-{Orn}-Pro-{dCha}-Trp-Arg (lactam bridge: Orn2-Arg6) | 219639-75-5 | MFCD07363866 | ≥98.0% | 896.09 g·mol⁻¹ | C47H65N11O7 | 25 MG
Supplier: Medchemexpress LLC HY10617825MG
PMX-53 is a synthetic peptidic antagonist of the complement C5a receptor (CD88) used in research to probe complement-mediated inflammation. It is orally active and inhibits C5a-induced neutrophil responses with a reported IC50 of ≈20 nM. The compound is supplied as a solid; molecular formula C47H65N11O7; molecular weight 896.09 g·mol⁻1; recommended storage at -20°C (in solvent: -80°C for long-term).
- Potent C5aR antagonist with low-nanomolar activity.
- Active in both in vitro and in vivo models.
- Peptidic structure suitable for mechanistic studies of complement signaling.
- Stable solid form with defined storage conditions.
- Available in small research pack sizes for experimental flexibility.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.