Specifications
| Certifications/Compliance | ANSI Z87.1 |
| Lens Coating | UV-treated |
| Certifications/Compliance (Europe) | EN-166 |
| Lens Tint | Clear |
| Frame Color | Black |
| Weight (Metric) | 25 g |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |