Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | ANSI Z87.1 |
| Description | Three lumens per side; The alkaline batteries provide up to 50 hours of use before replacing; Easy to use push button on/off switches arelocated at the top of each temple |
| Lens Tint | Clear |
| Magnification Power | +2.00 |
| Weight (Metric) | 50 g |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| Frame Type | Black frame with built-in swiveling LED bulbs |