Learn More
Cayman Chemical PncuronIumbromIde hydrate 10mg
SDP
Supplier:  Cayman Chemical 2377810 MG
Encompass_Preferred
An antagonist of muscle-type nAChRs (IC50 = 14.8 nM); acts as a non-depolarizing neuromuscular blocking agent; enhances isoflurane anesthesia (ED50 = 1.62 UG/kg to reduce immobilization)
Catalog No. 50-299-5053