Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pneumocandin B0 96.0+%, TCI America™
SDP

Catalog No. P253410MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P253410MG 10 mg
P253450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P253410MG Supplier TCI America Supplier No. P253410MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135575-42-7
Molecular Formula C50H80N8O17
Molecular Weight (g/mol) 1065.229
MDL Number MFCD28168025
InChI Key DQXPFAADCTZLNL-NRCXNPJQSA-N
PubChem CID 133562761
SMILES CCC(C)CC(C)CCCCCCCCC(=O)NC1CC(C(NC(=O)C2C(CCN2C(=O)C(NC(=O)C(NC(=O)C3CC(CN3C(=O)C(NC1=O)C(C)O)O)C(C(C4=CC=C(C=C4)O)O)O)C(CC(=O)N)O)O)O)O

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 1065.23
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pneumocandin B0
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.