Learn More
Promega pNL1.1.PGK[Nluc/PGK] Vector
Catalog No. PRN1441 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN1441 20 μg
1 options
Catalog No. PRN1441 Supplier Promega Supplier No. N1441
The promoter-driven NanoLuc (Nluc) control vectors can be used to co-transfect with experimental firefly luciferase vectors when using the Nano-Glo™ Dual-Luciferase Reporter (NanoDLR) Assay System.
  • Vectors Containing NanoLuc(R) Luciferase Expressed from Mammalian Promoters
  • Multiple promoter options (CMV, TK and PGK) are available to obtain appropriate levels of the control reporter in your experimental system
  • Co-transfect with experimental firefly luciferase vectors as normalization control
  • Measure signal using the Nano-Glo(R) Dual-Luciferase(R) Reporter (NanoDLR™) Assay System
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C