Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pNL3.2[NlucP/minP] Vector

Catalog No. PRN1041 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1041 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN1041 Supplier Promega Supplier No. N1041
May include imposed supplier surcharges.
Add to Cart
Add to Cart

NanoLuc (Nluc) luciferase is a small enzyme (19.1kDa) engineered for optimal performance as a luminescent reporter. The Vectors offer a minimal promoter for cloning response elements of interest.

  • Clone Response Elements of Interest to Drive Expression of NanoLuc(R) Luciferase
  • Nluc genes are codon optimized for mammalian cells
  • Many potential regulatory elements or other undesirable features have been removed (such as common restriction enzyme sites)
  • Vectors have a pGL4-based backbone for easy sequence transfer from existing plasmids

Specifications

Quantity 20 μg
Content And Storage -30°C to -10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.