Learn More
Promega pNLCoI2[luc2-P2A-NlucP/minP/Hygro] Vector
Catalog No. PRN1471 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN1471 20 μg
1 options
Catalog No. PRN1471 Supplier Promega Supplier No. N1471
The pNLCoI Vectors comprise a second-generation coincidence reporter vector system that allow expression of both firefly luciferase (luc2) and NanoLuc Luciferase fused to a PEST destabilization domain (NlucP) from the same mRNA transcript.
  • Vectors That Express Both Firefly and NanoLuc(R) Luciferases from the Same Transcript
  • Firefly and NanoLuc(R) luciferase have dissimilar compound interference profiles for better identifying more false-positives versus individually
  • Using two different transcriptional reporters reduces false hit rates and increases the identification of true biological hits
  • luc2 and NlucP provide a bright reporter combination compatible with low-copy-number and plate scale-up, and provide greater signal-to-background ratios
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C