Learn More
Promega pNLF1-secN [CMV/Hygro] Vector
Catalog No. PRN1371 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
20 μg
Catalog No. Quantity
PRN1371 20 μg
1 options
Catalog No. PRN1371 Supplier Promega Supplier No. N1371
Generate N-Terminal Fusions with N-Terminal Secreted NanoLuc™ Luciferase. The pNLF1 Vectors use traditional cloning with a multiple cloning site to generate N-terminal fusions to NanoLuc™ luciferase. All vectors contain a mammalian selectable marker to create a stable line.
  • Generate N- or C-Terminal Fusions with NanoLuc(R) Enzyme or N-Terminal Secreted NanoLuc(R) Luciferase
  • Use traditional cloning to insert protein of interest into the multiple cloning site (MCS)
  • Bright NanoLuc(R) reporter enables endogenous expression levels of fusion proteins for biologically relevant results
  • Choose from transient transfection or select for stable expression using hygromycin
Specifications
Quantity 20 μg
Content And Storage -30°C to -10°C