Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PNLIPRP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110186
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110186 Supplier Invitrogen™ Supplier No. PA5110186
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PNLIPRP1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot

PLRP2 may function as inhibitor of dietary triglyceride digestion. Lacks detectable lipase activity towards triglycerides, diglycerides, phosphatidylcholine, galactolipids or cholesterol esters (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PNLIPRP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PNLIPRP1
Gene Accession No. P54315, P54316, Q5BKQ4
Gene Alias inactive pancreatic lipase-related protein 1; pancreatic lipase related protein 1; pancreatic lipase-related protein 1; Plrp1; PL-RP1; Pnliprp1
Gene Symbols PNLIPRP1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18946, 5407, 84028
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.